MilliporeSigma
  • Differential effects of dolutegravir, bictegravir and raltegravir in adipokines and inflammation markers on human adipocytes.
Differential effects of dolutegravir, bictegravir and raltegravir in adipokines and inflammation markers on human adipocytes.
Life sciences (2022-09-13)
Pere Domingo, Tania Quesada-López, Joan Villarroya, Montserrat Cairó, Maria Del Mar Gutierrez, Maria Gracia Mateo, Isabel Mur, Noemí Corbacho, Joan Carles Domingo, Francesc Villarroya, Marta Giralt
ABSTRACT
To assess the potential direct effects of the integrase strand-transfer inhibitors (INsTIs) dolutegravir, bictegravir, and raltegravir, drugs used as treatment for people living with human immunodeficiency virus (PLWH), on human adipose cells. Drugs were added to the differentiation medium of human Simpson-Golabi-Behmel syndrome (SGBS) adipose cells and morphological adipogenesis was monitored for 10 days. Also, adipocytes were exposed to drugs following differentiation (day 14). The gene expression levels of selected adipogenesis markers, adipocyte metabolism markers, adipokines, and cytokines were determined by quantitative-reverse transcription polymerase-chain reaction. The release of adiponectin and leptin into the culture medium was measured using specific enzyme-linked immunosorbent assay, and release of interleukin-6 and chemokine (CC motif) ligand-2 using Multiplex assays. Overall morphological adipogenesis was unaltered by INsTIs. The expression of adipogenesis marker genes (peroxisome proliferator-activated receptor-Ɣ and lipoprotein lipase) was slightly reduced in dolutegravir-treated differentiating adipocytes. Bictegravir repressed gene expression and the release of pro-inflammatory cytokines in differentiating adipocytes. Dolutegravir and raltegravir increased interleukin-6 gene expression, but only dolutegravir increased interleukin-6 release. Dolutegravir repressed adiponectin expression and release in differentiating adipocytes and had a similar but milder effect on leptin. Drug treatment of mature adipocytes reduced adiponectin gene expression in response to dolutegravir. The INsTIs studied do not have a significant effect on human adipose cell differentiation but exert distinct effects on gene expression and secretion of adipokines and cytokines. These findings will help understand and manage the effects of INsTI-containing treatments on body weight and metabolic dysregulation in PLWH.
Sigma-Aldrich
Raltegravir, ≥98% (HPLC)