MilliporeSigma
Investigational therapies for the treatment of narcolepsy.
Expert opinion on investigational drugs (2017-07-21)
Stefano de Biase, Annacarmen Nilo, Gian Luigi Gigli, Mariarosaria Valente
ABSTRACT
Narcolepsy is a chronic sleep disorder characterized by a pentad of excessive daytime sleepiness (EDS), cataplexy, sleep paralysis, hypnagogic/hypnopompic hallucinations, and disturbed nocturnal sleep. While non-pharmacological treatments are sometimes helpful, more than 90% of narcoleptic patients require a pharmacological treatment. Areas covered: The present review is based on an extensive Internet and PubMed search from 1994 to 2017. It is focused on drugs currently in development for the treatment of narcolepsy. Expert opinion: Currently there is no cure for narcolepsy, with treatment focusing on symptoms control. However, these symptomatic treatments are often unsatisfactory. The research is leading to a better understanding of narcolepsy and its symptoms. New classes of compounds with possible applications in the development of novel stimulant/anticataplectic medications are described. H3 receptor antagonists represent a new therapeutic option for EDS in narcolepsy. JZP-110, with its distinct mechanism of action, would be a new therapeutic option for the treatment of EDS in the coming years. In the future, hypocretin-based therapies and immune-based therapies, could modify the clinical course of the disease. However, more information would be necessary to completely understand the autoimmune process and also how this process can be altered for therapeutic benefits.
Sigma-Aldrich
Solriamfetol hydrochloride, ≥98% (HPLC)