MilliporeSigma
  • Production of catalase and glucose oxidase by Aspergillus niger using unconventional oxygenation of culture.
Production of catalase and glucose oxidase by Aspergillus niger using unconventional oxygenation of culture.
Journal of applied microbiology (2000-08-17)
J Fiedurek, A Gromada
ABSTRACT
A novel method for increasing dissolved oxygen concentration in culture media has been developed. It involves adding hydrogen peroxide (H2O2) to the medium which is then decomposed to oxygen and water by catalase. Some factors affecting oxygenation of culture were investigated. Maximal oxygen concentration occurred in 50 ml of the medium containing 0-2 g wet mycelium and 0.2% glucose at pH 5.0. A new apparatus for automated addition of H2O2 to the bioreactor to keep the dissolved oxygen concentration constant over the range 1-100% +/- 2% was tested. A significant increase (over sixfold) of intracellular catalase activity was obtained while the dissolved oxygen concentration remained stable (30% +/- 2%).
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein