MilliporeSigma
  • Negative Staining Electron Microscopy for Morpho-Functional Characterization of Purified Golgi Membranes from Mammalian Cells.
Negative Staining Electron Microscopy for Morpho-Functional Characterization of Purified Golgi Membranes from Mammalian Cells.
Methods in molecular biology (Clifton, N.J.) (2022-12-14)
Stefano De Tito, Gabriele Turacchio, Carmen Valente
ABSTRACT
The Golgi apparatus is a highly dynamic organelle that controls lipid and protein sorting in the endocytic and exocytic cellular pathways. Perturbation of the lipid homeostasis or of the molecular machineries that regulate membrane remodeling/trafficking events on the Golgi membranes can dramatically change the morphology and functions of the Golgi apparatus. So far, several approaches have been described to characterize and define the Golgi morphology in intact cells and in vitro. Here, we describe the application of negative staining (NS) electron microscopy (EM) on purified Golgi membranes from HeLa cells. This approach allows to quantify and functionally characterize membrane remodeling events upon specific treatments that alter the Golgi morphology.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, FITC conjugate, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-MAN1A2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution