MilliporeSigma
  • Regulator of G protein signaling 12 drives inflammatory arthritis by activating synovial fibroblasts through MYCBP2/KIF2A signaling.
Regulator of G protein signaling 12 drives inflammatory arthritis by activating synovial fibroblasts through MYCBP2/KIF2A signaling.
Molecular therapy. Nucleic acids (2023-01-27)
Gongsheng Yuan, Shu-Ting Yang, Shuying Yang
ABSTRACT
Synovial fibroblasts are the active and aggressive drivers in the progression of arthritis, but the cellular and molecular mechanisms remain unknown. Here, our results showed that regulator of G protein signaling 12 (RGS12) maintained ciliogenesis in synovial fibroblasts, which is critical for the development of inflammatory arthritis. Deletion of RGS12 led to a significant decrease in ciliogenesis, adhesion, migration, and secretion of synovial fibroblasts. Mechanistically, RGS12 overexpression in synovial fibroblasts increased the length and number of cilia but decreased the protein level of kinesin family member 2A (KIF2A). The results of LC-MS analyses showed that RGS12 interacted with MYC binding protein 2 to enhance its ubiquitination activity, through which the KIF2A protein was degraded in synovial fibroblasts. Moreover, overexpression of KIF2A blocked the increases in cilia length and number. Mice with RGS12 deficiency or treatment of RGS12 shRNA nanoparticles significantly decreased the clinical score, paw swelling, synovitis, and cartilage destruction during inflammatory arthritis by inhibiting the activation of synovial fibroblasts. Therefore, this study provides evidence that RGS12 activates synovial fibroblasts' pathological function via promoting MCYBP2-mediated degradation of KIF2A and ciliogenesis. Our data suggest that RGS12 may be a potential drug target for the treatment of inflammatory arthritis.
Sigma-Aldrich
Anti-MYCBP2 Antibody, clone PM-8A9, clone PM-8A9, from mouse
Sigma-Aldrich
Anti-RGS12 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
C20A4 Human Chondrocyte Cell Line, C20A4 Human Chondrocyte Cell Line is widely used as a model cell line for studying normal and pathological cartilage repair mechanisms related to chondrocyte biology and physiology.