MilliporeSigma
  • Oncostatin M Receptor as a Therapeutic Target for Radioimmune Therapy in Synovial Sarcoma.
Oncostatin M Receptor as a Therapeutic Target for Radioimmune Therapy in Synovial Sarcoma.
Pharmaceuticals (Basel, Switzerland) (2022-06-25)
Sarah McCollum, Austen Kalivas, Matthew Kirkham, Kaden Kunz, Jeffrey Okojie, Adriene Pavek, Jared Barrott
ABSTRACT
Synovial sarcoma (SS) is a pediatric muscle cancer that primarily affects adolescents and young adults and has few treatment options. Complicating the treatment of synovial sarcoma is the low mutational burden of SS. Inflammatory pathways have been identified as being upregulated in some SS, leading to the discovery of upregulated oncostatin M receptor (OSMR). It was found that OSMR is upregulated in SS by RNAseq analysis and quantitative PCR, highlighting its potential in the treatment of SS. Also, OSMR is upregulated in mouse models for synovial sarcoma as demonstrated by western blot and immunohistochemistry, and the protein is present in both primary and metastatic sites of disease. Using a radioimmune therapy drug model, targeted therapy was synthesized for use in OSMR expressing SS and it was demonstrated that this drug is stable, while capable of efficient OSMR binding and isotope capture. Finally, this antibody conjugate exhibited ideal pharmacokinetics and targeted sites of disease in our mouse model and was taken up in both primary and metastatic diseased tissue. This suggests OSMR as an ideal target for therapy and this radioimmune therapy provides a novel treatment option for a disease with few therapy choices.
Sigma-Aldrich
Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.