MilliporeSigma
  • A Mixture of Dietary Plant Sterols at Nutritional Relevant Serum Concentration Inhibits Extrinsic Pathway of Eryptosis Induced by Cigarette Smoke Extract.
A Mixture of Dietary Plant Sterols at Nutritional Relevant Serum Concentration Inhibits Extrinsic Pathway of Eryptosis Induced by Cigarette Smoke Extract.
International journal of molecular sciences (2023-01-22)
Ignazio Restivo, Alessandro Attanzio, Luisa Tesoriere, Mario Allegra, Guadalupe Garcia-Llatas, Antonio Cilla
ABSTRACT
Cell death program of red blood cells (RBCs), called eryptosis, is characterized by activation of caspases and scrambling of membrane phospholipids with externalization of phosphatidylserine (PS). Excessive eryptosis confers a procoagulant phenotype and is implicated in impairment of microcirculation and increased prothrombotic risk. It has recently been reported that cigarette smokers have high levels of circulating eryptotic erythrocytes, and a possible contribution of eryptosis to the vaso-occlusive complications associated to cigarette smoke has been postulated. In this study, we demonstrate how a mixture of plant sterols (MPtS) consisting of β-sitosterol, campesterol and stigmasterol, at serum concentration reached after ingestion of a drink enriched with plant sterols, inhibits eryptosis induced by cigarette smoke extract (CSE). Isolated RBCs were exposed for 4 h to CSE (10-20% v/v). When RBCs were co-treated with CSE in the presence of 22 µM MPtS, a significant reduction of the measured hallmarks of apoptotic death like assembly of the death-inducing signaling complex (DISC), PS outsourced, ceramide production, cleaved forms of caspase 8/caspase 3, and phosphorylated p38 MAPK, was evident. The new beneficial properties of plant sterols on CSE-induced eryptosis presented in this work open new perspectives to prevent the negative physio-pathological events caused by the eryptotic red blood cells circulating in smokers.
Sigma-Aldrich
Monoclonal Anti-Ceramide antibody produced in mouse, clone MID 15B4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Mouse IgM+IgG+IgA Antibody, F(ab′)2, FITC conjugate, Species Adsorbed, 0.5 mg/mL, Chemicon®
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, HRP conjugate, Chemicon®, from rabbit