MilliporeSigma
  • Hippo signaling activates hedgehog signaling by Taz-driven Gli3 processing.
Hippo signaling activates hedgehog signaling by Taz-driven Gli3 processing.
Cell regeneration (London, England) (2023-02-01)
Chao Tang, Jirong Wang, Minli Yao, Xing Ji, Wei Shi, Chengyun Xu, Ling-Hui Zeng, Ximei Wu
ABSTRACT
The overlapping roles of Hippo and Hedgehog signaling in biological functions and diseases prompt us to investigate their potential interactions. Activation of Hippo signaling enhances the transcriptional output of Hedgehog signaling, and the role of Hippo signaling in regulating Hedgehog signaling relies on the Hippo pathway key effector, Taz. Interestingly, Taz exhibits a gradient expression across the posterior-to-anterior of limb bud mesoderms, similar to Sonic hedgehog (Shh). Importantly, Taz drives PKA to phosphorylate Gli3, resulting in the Gli3 processing into its repressor and attenuation of Hedgehog signaling in the Shh-independent manner. Specifically, Taz deletion in mouse embryonic limb bud mesenchyme not only enhances the Hedgehog signaling but partially restores the phenotypes from Shh deletion in causing severe defects of anteroposterior patterning and digit number and identity. Together, these results uncover Taz-dependent Gli3 processing as a hitherto uncharacterized mechanism controlling Hedgehog signaling, highlighting its cross-regulation by Hippo signaling.
Sigma-Aldrich
Anti-Protein patched homolog 1 Antibody, from rabbit, purified by affinity chromatography