MilliporeSigma
  • Self-Assembled PDINH Supramolecular System for Photocatalysis under Visible Light.
Self-Assembled PDINH Supramolecular System for Photocatalysis under Visible Light.
Advanced materials (Deerfield Beach, Fla.) (2016-06-17)
Di Liu, Jun Wang, Xiaojuan Bai, Ruilong Zong, Yongfa Zhu
ABSTRACT
A self-assembled perylene-3,4,9,10-tetracarboxylic diimide(PDINH) supramolecular system consisting of all-organic PDINH molecule building blocks through non-covalent interactions works as a visible light photocatalyst with high activity.
Sigma-Aldrich
PTCDI