MilliporeSigma
  • Oxymatrine ameliorates myocardial injury by inhibiting oxidative stress and apoptosis via the Nrf2/HO-1 and JAK/STAT pathways in type 2 diabetic rats.
Oxymatrine ameliorates myocardial injury by inhibiting oxidative stress and apoptosis via the Nrf2/HO-1 and JAK/STAT pathways in type 2 diabetic rats.
BMC complementary medicine and therapies (2023-01-04)
Yongpan Huang, Bin He, Chong Song, Xian Long, Jianbin He, Yansong Huang, Lijing Liu
ABSTRACT
The necessity of increasing the efficiency of organ preservation has encouraged researchers to explore the mechanisms underlying diabetes-related myocardial injuries. This study intended to evaluate the protective effects of oxymatrine (OMT) in myocardial injury caused by type 2 diabetes mellitus. A model of diabetic rats was established to simulate type 2 diabetes mellitus using an intraperitoneal injection of a single dose of 65 mg/kg streptozotocin with a high-fat and high-cholesterol diet, and diabetic rats were subsequently treated with OMT (60, 120 mg/kg) by gavage for 8 weeks. Thereafter, diabetic rats demonstrated notable decreases in left ventricular systolic pressure (LVSP), ±dp/dtmax, and in the activities of glutathione peroxidase, superoxide dismutase, and catalase. Moreover, we found notable increases in left ventricular end-diastolic pressure, fasting blood glucose, and malondialdehyde, as well as changes in cell apoptosis and decreased expression levels of Nrf2, HO-1, tyrosine protein kinase JAK (JAK), and signal transducer and transcription activator (STAT). Treatment with OMT alleviated all of the measured parameters. Collectively, these findings suggest that activation of the Nrf2/HO-1 and inhibition of the JAK/STAT signaling are involved in mediating the cardioprotective effects of OMT and also highlight the benefits of OMT in ameliorating myocardial injury in diabetic rats.
Sigma-Aldrich
IL-6 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)