MilliporeSigma
  • Na+-independent, nifedipine-resistant rat afferent arteriolar Ca2+ responses to noradrenaline: possible role of TRPC channels.
Na+-independent, nifedipine-resistant rat afferent arteriolar Ca2+ responses to noradrenaline: possible role of TRPC channels.
Acta physiologica (Oxford, England) (2010-04-30)
M Salomonsson, T H Braunstein, N-H Holstein-Rathlou, L J Jensen
ABSTRACT
In rat afferent arterioles we investigated the role of Na(+) entry in noradrenaline (NA)-induced depolarization and voltage-dependent Ca(2+) entry together with the importance of the transient receptor potential channel (TRPC) subfamily for non-voltage-dependent Ca(2+) entry. R (340/380) Fura-2 fluorescence was used as an index for intracellular free Ca(2+) concentration ([Ca(2+)](i)). Immunofluorescence detected the expression of TRPC channels. TRPC 1, 3 and 6 were expressed in afferent arteriolar vascular smooth muscle cells. Under extracellular Na(+)-free (0 Na) conditions, the plateau response to NA was 115% of the baseline R(340/380) (control response 123%). However, as the R(340/380) baseline increased (7%) after 0 Na the plateau reached the same level as during control conditions. Similar responses were obtained after blockade of the Na(+)/Ca(2+) exchanger. The L-type blocker nifedipine reduced the plateau response to NA both under control (from 134% to 116% of baseline) and 0 Na conditions (from 112% to 103% of baseline). In the presence of nifedipine, the putative TRPC channel blockers SKF 96365 (30 μm) and Gd(3+) (100 μm) further reduced the plateau Ca(2+) responses to NA (from 117% to 102% and from 117% to 110% respectively). We found that Na(+) is not crucial for the NA-induced depolarization that mediates Ca(2+) entry via L-type channels. In addition, the results are consistent with the idea that TRPC1/3/6 Ca(2+) -permeable cation channels expressed in afferent arteriolar smooth muscle cells mediate Ca(2+) entry during NA stimulation.
Sigma-Aldrich
Anti-TRPC1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-TRPC5 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-TRPC6 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-TRPC3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder