MilliporeSigma
  • D-2-hydroxyglutarate dehydrogenase governs adult neural stem cell activation and promotes histone acetylation via ATP-citrate lyase.
D-2-hydroxyglutarate dehydrogenase governs adult neural stem cell activation and promotes histone acetylation via ATP-citrate lyase.
Cell reports (2023-02-02)
Yinghao Liu, Min Wang, Ye Guo, Lei Wang, Weixiang Guo
ABSTRACT
The generation of neurons from quiescent radial-glia-like neural stem cells (RGLs) in adult brain goes hand in hand with the modulation of cellular metabolism. However, it is still unclear how the exact metabolic program governs the balance between quiescent and activated RGLs. Here, we find that loss of mitochondrial D-2-hydroxyglutarate dehydrogenase (D2HGDH) leads to aberrant accumulation of D-2-hydroxyglutarate (D-2-HG) and impaired RGL activation. Mechanistically, accumulated D-2-HG bonds directly to ATP-citrate lyase and competitively inhibits its enzymatic activity, thereby reducing acetyl-CoA production and diminishing histone acetylation. However, administration of acetate restores the acetyl-CoA levels via acetyl-CoA synthetase-mediated catabolism and rescues the deficiencies in histone acetylation and RGL activation caused by loss of D2HGDH. Therefore, our findings define the role of cross talk between mitochondria and the nucleus via a mitochondrial metabolite, D-2-HG, the aberrant accumulation of which hinders the regulation of histone acetylation in RGL activation and attenuates continuous neurogenesis in adult mammalian brain.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
D-α-Hydroxyglutaric acid disodium salt, ≥95% (GC)
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)