MilliporeSigma
  • Impact of Primary RPE Cells in a Porcine Organotypic Co-Cultivation Model.
Impact of Primary RPE Cells in a Porcine Organotypic Co-Cultivation Model.
Biomolecules (2022-07-28)
Natalie Wagner, Armin Safaei, José Hurst, Pia A Vogt, H Burkhard Dick, Stephanie C Joachim, Sven Schnichels
ABSTRACT
The pathological events of age-related macular degeneration are characterized by degenerative processes involving the photoreceptor cells, retinal pigment epithelium (RPE), and the Bruch's membrane as well as choroidal alterations. To mimic in vivo interactions between photoreceptor cells and RPE cells ex vivo, complex models are required. Hence, the aim of this study was to establish a porcine organotypic co-cultivation model and enlighten the interactions of photoreceptor and RPE cells, with a special emphasis on potential neuroprotective effects. Porcine neuroretina explants were cultured with primary porcine RPE cells (ppRPE) or medium derived from these cells (=conditioned medium). Neuroretina explants cultured alone served as controls. After eight days, RT-qPCR and immunohistology were performed to analyze photoreceptors, synapses, macroglia, microglia, complement factors, and pro-inflammatory cytokines (e.g., IL1B, IL6, TNF) in the neuroretina samples. The presence of ppRPE cells preserved photoreceptors, whereas synaptical density was unaltered. Interestingly, on an immunohistological as well as on an mRNA level, microglia and complement factors were comparable in all groups. Increased IL6 levels were noted in ppRPE and conditioned medium samples, while TNF was only upregulated in the ppRPE group. IL1B was elevated in conditioned medium samples. In conclusion, a co-cultivation of ppRPE cells and neuroretina seem to have beneficial effects on the neuroretina, preserving photoreceptors and maintaining synaptic vesicles in vitro. This organotypic co-cultivation model can be used to investigate the complex interactions between the retina and RPE cells, gain further insight into neurodegenerative pathomechanisms occurring in retinal diseases, and evaluate potential therapeutics.
Sigma-Aldrich
Donkey Anti-Chicken IgY Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8), liquid, clone 7E3-1B8, Calbiochem®