MilliporeSigma
  • p15INK4B is an alternative marker of senescent tumor cells in colorectal cancer.
p15INK4B is an alternative marker of senescent tumor cells in colorectal cancer.
Heliyon (2023-02-15)
Soon Sang Park, Young-Kyoung Lee, So Hyun Park, Su Bin Lim, Yong Won Choi, Jun Sang Shin, Young Hwa Kim, Jang-Hee Kim, Tae Jun Park
ABSTRACT
Senescent tumor cells are nonproliferating tumor cells which are closely related to cancer progression by secreting senescence-related molecules, called senescence-associated secreting phenotypes. Therefore, the presence of senescent tumor cells is considered a prognostic factor in various cancer types. Although senescence-associated β-galactosidase staining is considered the best marker for detection of senescent tumor cells, it can only be performed in fresh-frozen tissues. p16INK4A, a cyclin-dependent inhibitor, has been used as an alternative marker to detect senescent tumor cells in formalin-fixed paraffin-embedded tissues. However, other reliable markers to detect senescent tumor cells is still lacking. In the present study, using public single-cell RNA-sequencing data, we found that p15INK4B, a cyclin-dependent kinase inhibitor, is a novel marker for detection of senescent tumor cells. Moreover, p15INK4B expression was positively correlated with that of p16INK4A in colorectal cancer tissues. In in vitro studies, mRNA expression of p15INK4B was increased together with that of p16INK4A in H2O2- and therapy-induced cancer senescence models. However, the mRNA level of p15INK4B did not increase in the oncogene-induced senescence model in primary colonic epithelial cells. In conclusion, p15INK4B is a potential alternative marker for detection of senescent tumor cells together with conventional markers in advanced stages of colorectal cancer.
Sigma-Aldrich
Anti-p15 INK antibody produced in rabbit, affinity isolated antibody