MilliporeSigma
Retromer dysfunction in amyotrophic lateral sclerosis.
Proceedings of the National Academy of Sciences of the United States of America (2022-06-25)
Eduardo J Pérez-Torres, Irina Utkina-Sosunova, Vartika Mishra, Peter Barbuti, Mariangels De Planell-Saguer, Georgia Dermentzaki, Heather Geiger, Anna O Basile, Nicolas Robine, Delphine Fagegaltier, Kristin A Politi, Paola Rinchetti, Vernice Jackson-Lewis, Matthew Harms, Hemali Phatnani, Francesco Lotti, Serge Przedborski
ABSTRACT
Retromer is a heteropentameric complex that plays a specialized role in endosomal protein sorting and trafficking. Here, we report a reduction in the retromer proteins-vacuolar protein sorting 35 (VPS35), VPS26A, and VPS29-in patients with amyotrophic lateral sclerosis (ALS) and in the ALS model provided by transgenic (Tg) mice expressing the mutant superoxide dismutase-1 G93A. These changes are accompanied by a reduction of levels of the α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor subunit GluA1, a proxy of retromer function, in spinal cords from Tg SOD1G93A mice. Correction of the retromer deficit by a viral vector expressing VPS35 exacerbates the paralytic phenotype in Tg SOD1G93A mice. Conversely, lowering Vps35 levels in Tg SOD1G93A mice ameliorates the disease phenotype. In light of these findings, we propose that mild alterations in retromer inversely modulate neurodegeneration propensity in ALS.
Sigma-Aldrich
Anti-SNX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GluR1-NT (NT) Antibody, clone RH95, clone RH95, from mouse
Sigma-Aldrich
Anti-VPS29, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat