MilliporeSigma
  • Molecular design to regulate the photophysical properties of multifunctional TADF emitters towards high-performance TADF-based OLEDs with EQEs up to 22.4% and small efficiency roll-offs.
Molecular design to regulate the photophysical properties of multifunctional TADF emitters towards high-performance TADF-based OLEDs with EQEs up to 22.4% and small efficiency roll-offs.
Chemical science (2018-04-21)
Ling Yu, Zhongbin Wu, Guohua Xie, Weixuan Zeng, Dongge Ma, Chuluo Yang
ABSTRACT
Simultaneously achieving high efficiency and low efficiency roll-off remains a big challenge for OLEDs based on thermally activated delayed fluorescence (TADF) emitters. To address this issue, we elaborately designed and synthesized a series of new emitters with both TADF and aggregation-induced emission (AIE) properties by introducing 9,9-dimethyl-9,10-dihydroacridine (DMAC) or 10H-phenoxazine (PXZ) as donor units into a quinoxaline framework. By tuning the electron-donating capability of the donor as well as the amount of donor unit, the photophysical properties of the TADF-AIE emitters can be systematically regulated, with emissions ranging from green to red. We demonstrated efficient doped OLEDs with a maximum EQE of 22.4%, a maximum current efficiency (CEmax) of 80.3 cd A-1 and a maximum power efficiency (PEmax) of 64.1 lm W-1 for the green device, and an EQEmax of 14.1%, a CEmax of 36.1 cd A-1 and a PEmax of 28.1 lm W-1 for the orange device. Remarkably, these orange devices rendered small roll-offs of 1.4% and 21.3% respectively at a luminance of 100 and 1000 cd m-2. Attributed to the unique TADF and AIE features, the non-doped devices perform outstandingly with an EQEmax of 12.0%, a CEmax of 41.2 cd A-1 and a PEmax of 45.4 lm W-1.
Sigma-Aldrich
4,4′-Bis(carbazol-9-yl)biphenyl, ≥99% (HPLC)