MilliporeSigma
  • Proteasome inhibition targets the KMT2A transcriptional complex in acute lymphoblastic leukemia.
Proteasome inhibition targets the KMT2A transcriptional complex in acute lymphoblastic leukemia.
Nature communications (2023-02-14)
Jennifer L Kamens, Stephanie Nance, Cary Koss, Beisi Xu, Anitria Cotton, Jeannie W Lam, Elizabeth A R Garfinkle, Pratima Nallagatla, Amelia M R Smith, Sharnise Mitchell, Jing Ma, Duane Currier, William C Wright, Kanisha Kavdia, Vishwajeeth R Pagala, Wonil Kim, LaShanale M Wallace, Ji-Hoon Cho, Yiping Fan, Aman Seth, Nathaniel Twarog, John K Choi, Esther A Obeng, Mark E Hatley, Monika L Metzger, Hiroto Inaba, Sima Jeha, Jeffrey E Rubnitz, Junmin Peng, Taosheng Chen, Anang A Shelat, R Kiplin Guy, Tanja A Gruber
ABSTRACT
Rearrangments in Histone-lysine-N-methyltransferase 2A (KMT2Ar) are associated with pediatric, adult and therapy-induced acute leukemias. Infants with KMT2Ar acute lymphoblastic leukemia (ALL) have a poor prognosis with an event-free-survival of 38%. Herein we evaluate 1116 FDA approved compounds in primary KMT2Ar infant ALL specimens and identify a sensitivity to proteasome inhibition. Upon exposure to this class of agents, cells demonstrate a depletion of histone H2B monoubiquitination (H2Bub1) and histone H3 lysine 79 dimethylation (H3K79me2) at KMT2A target genes in addition to a downregulation of the KMT2A gene expression signature, providing evidence that it targets the KMT2A transcriptional complex and alters the epigenome. A cohort of relapsed/refractory KMT2Ar patients treated with this approach on a compassionate basis had an overall response rate of 90%. In conclusion, we report on a high throughput drug screen in primary pediatric leukemia specimens whose results translate into clinically meaningful responses. This innovative treatment approach is now being evaluated in a multi-institutional upfront trial for infants with newly diagnosed ALL.
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, culture supernatant, clone A3S, Upstate®
Sigma-Aldrich
ChIPAb+ Ubiquityl-Histone H2B - ChIP Validated Antibody and Primer Set, clone 56, from mouse, purified by using protein G