MilliporeSigma
  • Narrow Bandpass and Efficient Semitransparent Organic Solar Cells Based on Bioinspired Spectrally Selective Electrodes.
Narrow Bandpass and Efficient Semitransparent Organic Solar Cells Based on Bioinspired Spectrally Selective Electrodes.
ACS nano (2020-05-10)
Shu Wang, Jingde Chen, Ling Li, Lijian Zuo, Tian-Yi Qu, Hao Ren, Yanqing Li, Alex K-Y Jen, Jian-Xin Tang
ABSTRACT
The visual aesthetic that involves color, brightness, and glossiness is of great importance for building integrated photovoltaics. Semitransparent organic solar cells (ST-OSCs) are thus considered as the most promising candidate due to their superiority in transparency and efficiency. However, the realization of high color purity with narrow bandpass transmitted light usually causes the severely suppressed transparency in ST-OSCs. Herein, we present a spectrally selective electrode (SSE) by imitating the integrating strategy of beetle cuticle for achieving narrow bandpass ST-OSCs with high efficiency and long-term stability. The proposed SSE allows for efficient light-selective passage, leading to tunable narrow bandpass transmitted light from violet to red. An optimized power conversion efficiency of 15.07% is achieved for colorful ST-OSCs, which exhibit color purity close to 100% and a peak transmittance approaching 30%. Long-term stability is also improved for ST-OSCs made with this SSE due to the light-rejecting and the moisture-blocking abilities. The realization of bright and colorful ST-OSCs also indicates the application potential of SSEs in light-emitting diodes, lasers, and photodetectors.
Sigma-Aldrich
PTCDI
Pricing and availability is not currently available.