MilliporeSigma
  • Tankyrase-mediated ADP-ribosylation is a regulator of TNF-induced death.
Tankyrase-mediated ADP-ribosylation is a regulator of TNF-induced death.
Science advances (2022-05-12)
Lin Liu, Jarrod J Sandow, Deena M Leslie Pedrioli, Andre L Samson, Natasha Silke, Tobias Kratina, Rebecca L Ambrose, Marcel Doerflinger, Zhaoqing Hu, Emma Morrish, Diep Chau, Andrew J Kueh, Cheree Fitzibbon, Marc Pellegrini, Jaclyn S Pearson, Michael O Hottiger, Andrew I Webb, Najoua Lalaoui, John Silke
ABSTRACT
Tumor necrosis factor (TNF) is a key component of the innate immune response. Upon binding to its receptor, TNFR1, it promotes production of other cytokines via a membrane-bound complex 1 or induces cell death via a cytosolic complex 2. To understand how TNF-induced cell death is regulated, we performed mass spectrometry of complex 2 and identified tankyrase-1 as a native component that, upon a death stimulus, mediates complex 2 poly-ADP-ribosylation (PARylation). PARylation promotes recruitment of the E3 ligase RNF146, resulting in proteasomal degradation of complex 2, thereby limiting cell death. Expression of the ADP-ribose-binding/hydrolyzing severe acute respiratory syndrome coronavirus 2 macrodomain sensitizes cells to TNF-induced death via abolishing complex 2 PARylation. This suggests that disruption of ADP-ribosylation during an infection can prime a cell to retaliate with an inflammatory cell death.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Poly ADP-ribose Antibody, clone 10H, clone 10H, from mouse