MilliporeSigma
  • Quantitative phenotypic screens of Aspergillus niger mutants in solid and liquid culture.
Quantitative phenotypic screens of Aspergillus niger mutants in solid and liquid culture.
STAR protocols (2023-01-04)
Timothy Cairns, Xiaomei Zheng, Claudia Feurstein, Ping Zheng, Jibin Sun, Vera Meyer
ABSTRACT
This protocol describes procedures for quantifying Aspergillus niger growth in both solid and liquid culture. Firstly, by comparing radial growth between mutant and progenitor isolates on solid agar supplemented with sublethal stressors, susceptibility coefficients can be calculated. Secondly, analysis of macromorphological growth types in liquid culture allows full quantification of how a gene of interest affects submerged growth. By combining these assays, an extensive and quantitative dataset of how a gene of interest impacts growth in this fungus is possible. For complete details on the use and execution of this protocol, please refer to Cairns et al. (2019)1 and Cairns et al. (2022).2.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Caspofungin diacetate, ≥97% (HPLC)