MilliporeSigma
  • DNA damage triggers squamous metaplasia in human lung and mammary cells via mitotic checkpoints.
DNA damage triggers squamous metaplasia in human lung and mammary cells via mitotic checkpoints.
Cell death discovery (2023-01-22)
Lucía San Juan, Ana Freije, Natalia Sanz-Gómez, Beatriz Jiménez-Matías, Cayetano Pleguezuelos-Manzano, J Ramón Sanz, Ernesto de Diego, Sara Naranjo, Hans Clevers, Alberto Gandarillas
ABSTRACT
Epithelial transdifferentiation is frequent in tissue hyperplasia and contributes to disease in various degrees. Squamous metaplasia (SQM) precedes epidermoid lung cancer, an aggressive and frequent malignancy, but it is rare in the epithelium of the mammary gland. The mechanisms leading to SQM in the lung have been very poorly investigated. We have studied this issue on human freshly isolated cells and organoids. Here we show that human lung or mammary cells strikingly undergo SQM with polyploidisation when they are exposed to genotoxic or mitotic drugs, such as Doxorubicin or the cigarette carcinogen DMBA, Nocodazole, Taxol or inhibitors of Aurora-B kinase or Polo-like kinase. To note, the epidermoid response was attenuated when DNA repair was enhanced by Enoxacin or when mitotic checkpoints where abrogated by inhibition of Chk1 and Chk2. The results show that DNA damage has the potential to drive SQM via mitotic checkpoints, thus providing novel molecular candidate targets to tackle lung SCC. Our findings might also explain why SCC is frequent in the lung, but not in the mammary gland and why chemotherapy often causes complicating skin toxicity.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-Cdk1 Antibody, clone A17.1.1, clone A17.1.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vinculin Mouse mAb (V284 or VLN01), liquid, Calbiochem®, from mouse
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder