MilliporeSigma
  • An integrated model for Gpr124 function in Wnt7a/b signaling among vertebrates.
An integrated model for Gpr124 function in Wnt7a/b signaling among vertebrates.
Cell reports (2022-06-02)
Michelle America, Naguissa Bostaille, Marie Eubelen, Maud Martin, Didier Y R Stainier, Benoit Vanhollebeke
ABSTRACT
Within the central nervous system, Wnt7a/b are unambiguously discriminated from other Wnt ligands by an endothelial receptor complex made of the glycosylphosphatidylinositol (GPI)-anchored Reck and the adhesion G protein-coupled receptor (GPCR) Gpr124. Reck is a Wnt7a/b-specific receptor, while Gpr124 facilitates the delivery of Reck-bound Wnt7a/b ligands to Frizzled, through partially characterized mechanisms. We report that, in zebrafish, the Gpr124-Frizzled interactions are dominated by intracellular scaffolds that exploit the striking molecular mimicry between Gpr124 and Frizzled intracellular domains (ICDs): an internal Dvl-binding motif and a C-terminal ETTV motif that recruits Dlg4 and Magi3. By contrast, mammalian Gpr124 receptors exhibit an ICD-independent interaction mechanism governed by species-specific attributes of their transmembrane and extracellular domains. This mechanism seemingly evolved to replace the Dvl-mediated mechanism. By contrasting zebrafish, mouse, and human Gpr124, this study provides insights into the evolution of Gpr124/Reck function across the vertebrate clade, a receptor complex uniquely implicated in Wnt ligand-specific cellular responses.
Product Description
Roche
T7 RNA Polymerase, from Escherichia coli BL 21/pAR 1219
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep