MilliporeSigma
  • Enhanced Zika virus-like particle development using Baculovirus spp. constructs.
Enhanced Zika virus-like particle development using Baculovirus spp. constructs.
Journal of medical virology (2022-10-23)
Alexander Malogolovkin, Andrew Davies, Sherif Abouelhadid, Adeline Kerviel, Polly Roy, Andrew K Falconar
ABSTRACT
Zika virus (ZIKV) is one of several examples of an unprecedented pandemic spread and against which there is currently no suitable vaccine or treatment. Here, we constructed and characterized recombinant baculovirus-derived ZIKV-like particles (Zika VLPs) to study ZIKV-antibody interactions. These VLPs, uniquely consisted of the full-length ZIKV capsid (C), pre-membrane (prM), and envelope (E) proteins with either: a) the viral nonstructural NS2B and NS3 protease unit under one or two different promoters or b) an alternative host-cell furin protease encoding cleavage sequence inserted between the C and prM genes, together with lobster tropomyosin leader and honeybee signal sequences with one promoter for increased extracellular secretion. All these Zika VLPs displayed typical virion morphology in transmission electron microscopic analysis when expressed in both insect (Sf9) and mammalian (HEK293T) cells and no uncleaved prM glycoprotein was detected, as are present on immature virions. The importance of glycosylation of the E glycoprotein was shown by the effects on both polyclonal and monoclonal antibody reactions after these N-linked carbohydrate residues were disrupted by oxidation or enzymatic cleavage. Importantly, the construct which contained the host-cell furin protease cleavage sequence together with a lobster tropomyosin leader and honeybee signal sequences under one promoter produced higher Zika VLP titers and protein concentrations and which can now be tested as a superior construct in multifunctional diagnostic (ELISA and neutralization/antibody-dependent enhancement) assays and immunogenic assessments possibly leading to vaccine trials.
Sigma-Aldrich
Anti-JMJD1A Antibody, from rabbit, purified by affinity chromatography
Supelco
SYPRO® Ruby Protein Gel Stain