MilliporeSigma
  • A systemic cell stress signal confers neuronal resilience toward oxidative stress in a Hedgehog-dependent manner.
A systemic cell stress signal confers neuronal resilience toward oxidative stress in a Hedgehog-dependent manner.
Cell reports (2022-10-20)
Kyung Min Chung, Hyunha Kim, Cláudio Gouveia Roque, Ethan P McCurdy, Trang T T Nguyen, Markus D Siegelin, Jee-Yeon Hwang, Ulrich Hengst
ABSTRACT
Cells possess several conserved adaptive mechanisms to respond to stress. Stress signaling is initiated to reestablish cellular homeostasis, but its effects on the tissue or systemic levels are far less understood. We report that the secreted luminal domain of the endoplasmic reticulum (ER) stress transducer CREB3L2 (which we name TAILS [transmissible activator of increased cell livability under stress]) is an endogenous, cell non-autonomous activator of neuronal resilience. In response to oxidative insults, neurons secrete TAILS, which potentiates hedgehog signaling through direct interaction with Sonic hedgehog (SHH) and its receptor PTCH1, leading to improved antioxidant signaling and mitochondrial function in neighboring neurons. In an in vivo model of ischemic brain injury, administration of TAILS enables survival of CNS neurons and fully preserves cognitive function in behavioral tests. Our findings reveal an SHH-mediated, cell non-autonomous branch of cellular stress signaling that confers resilience to oxidative stress in the mature brain, providing protection from ischemic neurodegeneration.
Sigma-Aldrich
Anti-Tom20/Tomm20 Antibody, clone 2F8.1, clone 2F8.1, from mouse
Sigma-Aldrich
Anti-CREB3L2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution