MilliporeSigma
Digested Ara h 1 has sensitizing capacity in Brown Norway rats.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2009-08-20)
K L Bøgh, S Kroghsbo, L Dahl, N M Rigby, V Barkholt, E N C Mills, C B Madsen
ABSTRACT
Food allergies are a public health issue of growing concern, with peanuts in particular being associated with severe reactions. The peanut allergen, Ara h 1, belongs to the cupin plant food allergen family, which, unlike other structural families, appears to be broken down rapidly following gastrointestinal digestion. Using Ara h 1 as a model allergen, the ability of digested protein to sensitize has been investigated. Ara h 1 was purified from whole roasted peanuts. Intact Ara h 1 was digested in an in vitro model, simulating the human gastrointestinal digestion process. Digestion products were analysed for peptide sizes and their ability to aggregate. Brown Norway (BN) rats, used as an animal model, were immunized with purified intact Ara h 1 or the gastrointestinal digestion products thereof. The sensitizing capacity was evaluated by analyses of specific antibody (IgG1, IgG2a and IgE) responses and ability to trigger mediator release of rat basophilic leukaemia (RBL)-2H3 cells. The present study showed that Ara h 1 was broken down, resulting in peptide fragments of sizes<2.0 kDa, of which approximately 50% was in aggregated complexes of Mr up to 20 kDa. Ara h 1 digesta were shown to have sensitizing capacity in BN rats, being capable of inducing specific IgG and IgE antibodies. The IgE response was functional, having the capacity to induce specific degranulation of RBL cells. From this study, it can be concluded that lability of a food allergen to gastrointestinal digestion does not necessarily abrogate its allergenic sensitizing potential.
Sigma-Aldrich
Trypsin Agarose, buffered aqueous suspension, from bovine pancreas trypsin
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O