MilliporeSigma
  • Thyroid stimulating hormone aggravates diabetic retinopathy through the mitochondrial apoptotic pathway.
Thyroid stimulating hormone aggravates diabetic retinopathy through the mitochondrial apoptotic pathway.
Journal of cellular physiology (2021-08-27)
Dong Lin, Ruijie Qin, Lixin Guo
ABSTRACT
Diabetic retinopathy (DR) is a common complication of diabetes mellitus. High glucose-induced mitochondrial apoptosis is involved in the loss of retinal pericytes (PCs), which is considered to be a predominant pathologic change of diabetic retinopathy (DR). A high thyroid stimulating hormone (TSH) serum level is associated with an increased prevalence of DR in diabetic patients. Here, we investigated whether TSH regulated glucose-induced PCs loss through TSH-receptor (TSHR)-dependent mitochondrial apoptosis. First, the serum TSH level was found to be an independent risk factor for DR in Type 2 diabetic study participants (odds ratio = 2.294; 95% confidence interval: 1.925-2.733; p ≤ 0.001). Second, human PCs were treated with different concentrations of glucose, with or without bovine TSH (b-TSH). Glucose induced mitochondrial apoptosis through various mechanisms, including through regulating the expression of apoptosis-related proteins and inducing mitochondrial dysfunction, which could be deteriorated by costimulation of glucose and b-TSH. Additionally, we detected functional TSHR in PCs; blocking TSHR significantly restricted TSH-induced apoptosis. Thus, the presence of functional TSHR in human retinal PCs may facilitate the effect of high TSH on high glucose-induced PCs loss through TSHR-dependent mitochondrial apoptosis.
Sigma-Aldrich
Thyrotropic hormone from bovine pituitary