MilliporeSigma
  • Purified anacardic acids exert multiple neuroprotective effects in pesticide model of Parkinson's disease: in vivo and in silico analysis.
Purified anacardic acids exert multiple neuroprotective effects in pesticide model of Parkinson's disease: in vivo and in silico analysis.
IUBMB life (2020-05-26)
Ricielle L Augusto, Ingrid P Mendonça, Gabriel N de Albuquerque Rego, Danielle D Pereira, Lílian V da Penha Gonçalves, Maria L Dos Santos, Raphael F de Souza, Giselle M M Moreno, Pablo R G Cardoso, Daniele de Souza Andrade, José C da Silva-Júnior, Michelly C Pereira, Christina A Peixoto, Cybelle F B Medeiros-Linard, Ivone A de Souza, Belmira L da Silveira Andrade-da-Costa
ABSTRACT
Parkinson's disease (PD) induced by environmental toxins involves a multifactorial cascade of harmful factors, thus motivating the search for therapeutic agents able to act on the greatest number of molecular targets. This study evaluated the efficacy of 50 mg/kg purified anacardic acids (AAs), isolated from cashew nut shell liquid, on multiple steps of oxidative stress and inflammation induced by rotenone in the substantia nigra (SN) and striatum. Adult mice were divided into four groups: Control, rotenone, AAs + rotenone, and AAs alone. Lipoperoxidation, nitric oxide (NO) levels, and reduced glutathione (GSH)/oxidized gluthatione (GSSG) ratio were evaluated. NF-kB-p65, pro-IL-1β, cleaved IL-1β, metalloproteinase-9, Tissue Inhibitory Factor-1 (TIMP-1), tyrosine hydroxylase (TH), and glial fibrillary acidic protein (GFAP) levels were assessed by Western blot. In silico studies were also made using the SwissADME web tool. Rotenone increased lipoperoxidation and NO production and reduced TH levels and GSH/GSSG ratio in both SN and striatum. It also enhanced NF-kB-p65, pro, and cleaved IL-1β, MMP-9, GFAP levels compared to control and AAs groups. The AAs alone reduced pro-IL-1β in the striatum while they augmented TIMP1 and reduced MMP-9 amounts in both regions. AAs reversed rotenone-induced effects on lipoperoxidation, NO production, and GSH/GSSG ratio, as well as increased TH and attenuated pro-IL-1β and MMP-9 levels in both regions, NF-kB-p65 in the SN and GFAP in the striatum. Altogether, the in vivo and in silico analysis reinforced multiple and defined molecular targets of AAs, identifying that they are promising neuroprotective drug candidates for PD, acting against oxidative and inflammatory conditions induced by rotenone.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-NFKB p65 antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM273, purified immunoglobulin