MilliporeSigma
  • CCDC85B promotes non-small cell lung cancer cell proliferation and invasion.
CCDC85B promotes non-small cell lung cancer cell proliferation and invasion.
Molecular carcinogenesis (2018-09-23)
Yangyang Feng, Yue Gao, Juanhan Yu, Guiyang Jiang, Xiupeng Zhang, Xuyong Lin, Qiang Han, Xuezhu Rong, Hongtao Xu, Qingchang Li, Xueshan Qiu, Enhua Wang
ABSTRACT
Coiled-coil domain containing 85 B (CCDC85B) is involved in diverse biological processes; however, its expression patterns and functions in human cancers are yet unknown. The present study demonstrated that the expression of CCDC85B in the cytoplasm of the non-small cell lung cancer (NSCLC) tumor cells was significantly higher compared to adjacent normal lung tissues (P < 0.05). Furthermore, CCDC85B expression correlated with advanced TNM stage (P = 0.004) and positive regional lymph node metastasis (P = 0.009) of NSCLC. In addition, in A549 and H1299 lung cancer cell lines, the overexpression of CCDC85B promoted cell proliferation and invasion, while siRNA-mediated CCDC85B knockdown exhibited opposite effects. CCDC85B promoted AKT and GSK3β phosphorylation and upregulated the levels of active β-catenin, Wnt targets c-myc, cyclin D1, and MMP7. Besides, the CCDC85B-induced upregulation of phosphorylated GSK3β and active β-catenin was rescued following the treatment with PI3 K inhibitor, LY294002. In conclusion, CCDC85B was associated with NSCLC progression as it promoted the proliferation and invasion of lung cancer cells through activated AKT/GSK3β/β-catenin oncogenic signaling pathway. Therefore, CCDC85B might serve as a novel target for NSCLC treatment.
Sigma-Aldrich
Anti-CCDC85B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution