MilliporeSigma
  • KIBRA regulates activity-induced AMPA receptor expression and synaptic plasticity in an age-dependent manner.
KIBRA regulates activity-induced AMPA receptor expression and synaptic plasticity in an age-dependent manner.
iScience (2022-12-06)
Matthew L Mendoza, Lilyana D Quigley, Thomas Dunham, Lenora J Volk
ABSTRACT
A growing body of human literature implicates KIBRA in memory and neurodevelopmental disorders. Memory and the cellular substrates supporting adaptive cognition change across development. Using an inducible KIBRA knockout mouse, we demonstrate that adult-onset deletion of KIBRA in forebrain neurons impairs long-term spatial memory and long-term potentiation (LTP). These LTP deficits correlate with adult-selective decreases in extrasynaptic AMPA receptors under basal conditions, and we identify a role for KIBRA in LTP-induced AMPAR upregulation. In contrast, juvenile-onset deletion of KIBRA in forebrain neurons did not affect LTP and had minimal effects on basal AMPAR expression. LTP did not increase AMPAR protein expression in juvenile WT mice, providing a potential explanation for juvenile resilience to KIBRA deletion. These data suggest that KIBRA serves a unique role in adult hippocampal function through regulation of basal and activity-dependent AMPAR proteostasis that supports synaptic plasticity.
Sigma-Aldrich
Anti-PKC ζ antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-WWC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography