MilliporeSigma
  • Heparanase Increases Podocyte Survival and Autophagic Flux after Adriamycin-Induced Injury.
Heparanase Increases Podocyte Survival and Autophagic Flux after Adriamycin-Induced Injury.
International journal of molecular sciences (2022-10-28)
Hanan Abu-Tayeh Suleiman, Shereen Said, Haya Ali Saleh, Aviva Gamliel-Lazarovich, Eyas Haddad, Irina Minkov, Yaniv Zohar, Neta Ilan, Israel Vlodavsky, Zaid Abassi, Suheir Assady
ABSTRACT
The kidney glomerular filtration barrier (GFB) is enriched with heparan sulfate (HS) proteoglycans, which contribute to its permselectivity. The endoglycosidase heparanase cleaves HS and hence appears to be involved in the pathogenesis of kidney injury and glomerulonephritis. We have recently reported, nonetheless, that heparanase overexpression preserved glomerular structure and kidney function in an experimental model of Adriamycin-induced nephropathy. To elucidate mechanisms underlying heparanase function in podocytes-key GFB cells, we utilized a human podocyte cell line and transgenic mice overexpressing heparanase. Notably, podocytes overexpressing heparanase (H) demonstrated significantly higher survival rates and viability after exposure to Adriamycin or hydrogen peroxide, compared with mock-infected (V) podocytes. Immunofluorescence staining of kidney cryo-sections and cultured H and V podocytes as well as immunoblotting of proteins extracted from cultured cells, revealed that exposure to toxic injury resulted in a significant increase in autophagic flux in H podocytes, which was reversed by the heparanase inhibitor, Roneparstat (SST0001). Heparanase overexpression was also associated with substantial transcriptional upregulation of autophagy genes BCN1, ATG5, and ATG12, following Adriamycin treatment. Moreover, cleaved caspase-3 was attenuated in H podocytes exposed to Adriamycin, indicating lower apoptotic cell death in H vs. V podocytes. Collectively, these findings suggest that in podocytes, elevated levels of heparanase promote cytoprotection.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Anti-LC3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho Beclin Antibody (Thr119), from rabbit, purified by affinity chromatography