MilliporeSigma
  • GM-CSF (granulocyte-macrophage colony-stimulating factor) treatment improves sperm parameters in men with oligoasthenoteratospermia via PI3K/AKT pathway.
GM-CSF (granulocyte-macrophage colony-stimulating factor) treatment improves sperm parameters in men with oligoasthenoteratospermia via PI3K/AKT pathway.
Andrologia (2022-05-12)
Fatemeh Tanhaye Kalate Sabz, Fatemeh Sadat Amjadi, Zahra Zandieh, Elham Hosseini, Reza Aflatoonian, Maryam Tabatabaei, Masoud Mohammadian, Mahnaz Ashrafi
ABSTRACT
Poor sperm quality in oligoasthenoteratospermia patients negatively affects assisted reproductive technology outcomes. Therefore, the development of sperm media is necessary to improve sperm parameters. This study investigated the effect of GM-CSF via PI3K/AKT pathway on sperm quality in OAT patients. Semen samples were collected from 20 OAT patients, and each sample was divided into two groups: Experiment and Control. In the experimental group, the samples were incubated with medium containing GM-CSF, and control samples were incubated without GM-CSF. Sperm parameters, mitochondrial membrane potential, acrosome reaction and DFI were studied; in addition, gene expression of PI3KR1, PI3KCA, GLUT1, GLUT3 and AKT1 was analysed, evaluation of PAKT/TAKT, and expression of GLUT 1, 3 was examined; subsequent fertilization rate and embryo quality were assessed. Our data showed that GM-CSF supplementation could significantly increase motility, mitochondrial activity, gene expression of PI3KCA, AKT1, the protein level of PAKT/TAKT and expression of GLUT 1, 3 while it decreases DNA fragmentation. The fertilization rate and embryo quality significantly improved in the treatment group. LY294002 had adverse effects on sperm motility and the PAKT/TAKT ratio. GM-CSF can improve in vitro sperm quality and could be a suitable supplement to sperm media for OAT patients.
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Lectin from Arachis hypogaea (peanut), FITC conjugate, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in E. coli, suitable for cell culture