MilliporeSigma
  • Imaging Platelets and Megakaryocytes by High-Resolution Laser Fluorescence Microscopy.
Imaging Platelets and Megakaryocytes by High-Resolution Laser Fluorescence Microscopy.
Methods in molecular biology (Clifton, N.J.) (2018-09-02)
Fred G Pluthero, Walter H A Kahr
ABSTRACT
Microscopy is central to studies of platelets and their precursor megakaryocytes. Here we describe methods to rapidly obtain high resolution images of fixed platelets, megakaryocytes and megakaryocytic cells via immunofluorescence microscopy. Protocols covered include: (1) isolation and preparation of cells suitable for fluorescence staining; (2) staining with antibodies and other molecules; (3) imaging via spinning-disc confocal and structured illumination laser fluorescence microscopy; (4) processing and presentation of images. Also included is a list of primary antibodies we have validated for use in staining specific proteins and subcellular structures in platelets and megakaryocytes.
Sigma-Aldrich
Anti-ARPC1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ETV6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-RAR alpha Antibody, clone 76, clone 763, Chemicon®, from mouse