MilliporeSigma
  • Proteomics Identifies Substrates and a Novel Component in hSnd2-Dependent ER Protein Targeting.
Proteomics Identifies Substrates and a Novel Component in hSnd2-Dependent ER Protein Targeting.
Cells (2022-09-24)
Andrea Tirincsi, Sarah O'Keefe, Duy Nguyen, Mark Sicking, Johanna Dudek, Friedrich Förster, Martin Jung, Drazena Hadzibeganovic, Volkhard Helms, Stephen High, Richard Zimmermann, Sven Lang
ABSTRACT
Importing proteins into the endoplasmic reticulum (ER) is essential for about 30% of the human proteome. It involves the targeting of precursor proteins to the ER and their insertion into or translocation across the ER membrane. Furthermore, it relies on signals in the precursor polypeptides and components, which read the signals and facilitate their targeting to a protein-conducting channel in the ER membrane, the Sec61 complex. Compared to the SRP- and TRC-dependent pathways, little is known about the SRP-independent/SND pathway. Our aim was to identify additional components and characterize the client spectrum of the human SND pathway. The established strategy of combining the depletion of the central hSnd2 component from HeLa cells with proteomic and differential protein abundance analysis was used. The SRP and TRC targeting pathways were analyzed in comparison. TMEM109 was characterized as hSnd3. Unlike SRP but similar to TRC, the SND clients are predominantly membrane proteins with N-terminal, central, or C-terminal targeting signals.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-TMEM109 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Roche
Nuclease S7, Micrococcal nuclease, from Staphylococcus aureus