MilliporeSigma
  • Inoculation of tannin-degrading bacteria into novel hosts increases performance on tannin-rich diets.
Inoculation of tannin-degrading bacteria into novel hosts increases performance on tannin-rich diets.
Environmental microbiology (2015-03-11)
Kevin D Kohl, Ashley Stengel, M Denise Dearing
ABSTRACT
It has been hypothesized that herbivores host tannin-degrading bacteria (TDB) to overcome the toxic challenges posed by plant tannins. While TDB have been isolated from the guts of numerous mammals, their functional significance to their hosts has never been explicitly tested. We introduced TDB into lab rats, which do not host TDB, and measured host performance on tannin-rich diets. We first isolated three species of TDB, Escherichia coli, Bacillus subtilis and Enterococcus faecalis, from the guts of the desert woodrat (Neotoma lepida), which regularly feeds on tannin-rich plants. Then, we inoculated isolated TDB, as well as full woodrat microbial communities into laboratory rats. A control group was inoculated with sterilized woodrat faeces. Recipient lab rats were fed increasing concentrations of tannic acid, and we monitored tannic acid intake, body mass and liver damage as measured by serum alanine aminotransferase activity. Lab rats given TDB as isolates or full communities exhibited increased tannic acid intake, higher maintenance of body mass and lower indicators of liver damage compared with control animals. These differences were maintained when the trial was repeated after 6 weeks of feeding on tannin-free diets. Our results are the first to demonstrate that TDB significantly increase host performance on tannin-rich diets.
Sigma-Aldrich
Tannase from Aspergillus ficuum, powder, white, ≥150 U/g