MilliporeSigma
  • Muscle and nerve net organization in stalked jellyfish (Medusozoa: Staurozoa).
Muscle and nerve net organization in stalked jellyfish (Medusozoa: Staurozoa).
Journal of morphology (2016-10-04)
Hannah E Westlake, Louise R Page
ABSTRACT
Staurozoan cnidarians display an unusual combination of polyp and medusa characteristics and their morphology may be informative about the evolutionary origin of medusae. We studied neuromuscular morphology of two staurozoans, Haliclystus 'sanjuanensis' and Manania handi, using whole mount immunohistochemistry with antibodies against FMRFamide and α-tubulin to label neurons and phalloidin to label muscles. All muscles appeared to lack striations. Longitudinal interradial muscles are probable homologues of stalk muscles in scyphopolyps, but in adult staurozoans they are elaborated to inwardly flex marginal lobes of the calyx during prey capture; these muscles are pennate in M. handi. Manubrial perradial muscles, like the manubrium itself, are an innovation shared with pelagic medusae and manubrial interradial muscles are shared with scyphozoan ephyra. Marginal muscles of M. handi displayed occasional synchronous contraction reminiscent of a medusa swim pulse, but contractions were not repetitive. The nerve net in both species showed regional variation in density and orientation of neurons. Some areas labeled predominantly by α-tubulin antibodies (exumbrellar epidermis), other areas labeled exclusively by FMRFamide antibodies (dense plexus of neurites surrounding the base of secondary tentacles, neuronal concentration at the base of transformed primary tentacles; gastrodermal nerve net), but most areas showed a mix of neurons labeled by these two antibodies and frequent co-labeling of neurons. Transformed primary tentacles had a concentration of FMRFamide-immunoreactive neurons at their base that was associated with a pigment spot in M. handi; this is consistent with their homology with rhopalia of medusae, which are also derived from primary tentacles. The muscular system of these staurozoans embodies characteristics of both scyphopolyps and pelagic medusae. However, their nerve net is more polyp-like, although marginal concentrations of the net associated with primary and secondary tentacles may facilitate the richer behavioral repertoire of staurozoans relative to polyps of other medusozoans. J. Morphol. 278:29-49, 2017. ©© 2016 Wiley Periodicals,Inc.