MilliporeSigma
  • Sphingosine kinase 2 is essential for remyelination following cuprizone intoxication.
Sphingosine kinase 2 is essential for remyelination following cuprizone intoxication.
Glia (2021-08-17)
Huitong Song, Holly P McEwen, Thomas Duncan, Jun Yup Lee, Jonathan D Teo, Anthony S Don
ABSTRACT
Therapeutics that promote oligodendrocyte survival and remyelination are needed to restore neurological function in demyelinating diseases. Sphingosine 1-phosphate (S1P) is an essential lipid metabolite that signals through five G-protein coupled receptors. S1P receptor agonists such as Fingolimod are valuable immunosuppressants used to treat multiple sclerosis, and promote oligodendrocyte survival. However, the role for endogenous S1P, synthesized by the enzyme sphingosine kinase 2 (SphK2), in oligodendrocyte survival and myelination has not been established. This study investigated the requirement for SphK2 in oligodendrocyte survival and remyelination using the cuprizone mouse model of acute demyelination, followed by spontaneous remyelination. Oligodendrocyte density did not differ between untreated wild-type (WT) and SphK2 knockout (SphK2-/- ) mice. However, cuprizone treatment caused significantly greater loss of mature oligodendrocytes in SphK2-/- compared to WT mice. Following cuprizone withdrawal, spontaneous remyelination occurred in WT but not SphK2-/- mice, even though progenitor and mature oligodendrocyte density increased in both genotypes. Levels of cytotoxic sphingosine and ceramide were higher in the corpus callosum of SphK2-/- mice, and in contrast to WT mice, did not decline following cuprizone withdrawal in SphK2-/- mice. We also observed a significant reduction in myelin thickness with aging in SphK2-/- compared to WT mice. These results provide the first evidence that SphK2, the dominant enzyme catalyzing S1P synthesis in the adult brain, is essential for remyelination following a demyelinating insult and myelin maintenance with aging. We propose that persistently high levels of sphingosine and ceramide, a direct consequence of SphK2 deficiency, may block remyelination.
Sigma-Aldrich
Anti-NG2 Antibody, clone 1L2, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial