MilliporeSigma
  • Development and validation of the maximal electro-shock seizure model in dogs.
Development and validation of the maximal electro-shock seizure model in dogs.
Journal of veterinary pharmacology and therapeutics (2007-11-10)
P R Territo, K J Freise, K Newhall, S D Barnhart, S C Peters, D R Engleking, T J Burnett, B Abdul-Karim, H E Shannon
ABSTRACT
The development and validation of the maximal electro-shock (MES) model using phenobarbital (Pb) as the positive control is described. This approach builds on previous work in rodent model systems, and has been adapted to dogs as a tool for pharmaceutical dose selection. Dogs, like rodents, exhibit generalized convulsions which manifest as progressive clinical signs in a dose (electrical current) dependent fashion. At the limit (300 mA, 200 msec) animals underwent clonic-tonic convulsions consistent with complete generalized (Grand Mal) seizures with a grade 3 clinical score (CS) and a menace response time of 98.5 +/- 24.4 sec (n = 8). Pretreatment of animals with Pb at 3, 10, and 30 mg/kg, in a 4-by-4 complete block crossover design (Latin-Square), resulted in a dose-dependant reduction in CS and menace response time. Estimates of plasma Pb concentration taken prior to MES induction showed a similar dose-dependent reduction in CS and menace response time with concentration. Using a cumulative logistic regression model, a predicted 50% probability of a CS = 1 was approximately 11.4 mg/kg. In addition, plasma Pb concentrations predicted a 50% probability of a CS = 1 occurs at plasma Pb concentration of approximately 16.0 mug/mL. Combined these data suggest that MES is a useful model for evaluating generalized convulsions in canines and may provide a tool for dose selection of novel pharmaceutical compounds.