MilliporeSigma
  • Subcellular reorganization and altered phosphorylation of the astrocytic gap junction protein connexin43 in human and experimental temporal lobe epilepsy.
Subcellular reorganization and altered phosphorylation of the astrocytic gap junction protein connexin43 in human and experimental temporal lobe epilepsy.
Glia (2017-08-11)
Tushar Deshpande, Tingsong Li, Michel K Herde, Albert Becker, Hartmut Vatter, Martin K Schwarz, Christian Henneberger, Christian Steinhäuser, Peter Bedner
ABSTRACT
Dysfunctional astrocytes are increasingly recognized as key players in the development and progression of mesial temporal lobe epilepsy (MTLE). One of the dramatic changes astrocytes undergo in MTLE with hippocampal sclerosis (HS) is loss of gap junction coupling. To further elucidate molecular mechanism(s) underlying this alteration, we assessed expression, cellular localization and phosphorylation status of astrocytic gap junction proteins in human and experimental MTLE-HS. In addition to conventional confocal analysis of immunohistochemical staining we employed expansion microscopy, which allowed visualization of blood-brain-barrier (BBB) associated cellular elements at a sub-µm scale. Western Blot analysis showed that plasma membrane expression of connexin43 (Cx43) and Cx30 were not significantly different in hippocampal specimens with and without sclerosis. However, we observed a pronounced subcellular redistribution of Cx43 toward perivascular endfeet in HS, an effect that was accompanied by increased plaque size. Furthermore, in HS Cx43 was characterized by enhanced C-terminal phosphorylation of sites affecting channel permeability. Prominent albumin immunoreactivity was found in the perivascular space of HS tissue, indicating that BBB damage and consequential albumin extravasation was involved in Cx43 dysregulation. Together, our results suggest that subcellular reorganization and/or abnormal posttranslational processing rather than transcriptional downregulation of astrocytic gap junction proteins account for the loss of coupling reported in human and experimental TLE. The observations of the present study provide new insights into pathological alterations of astrocytes in HS, which may aid in the identification of novel therapeutic targets and development of alternative anti-epileptogenic strategies.
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Chondroitin Sulfate Proteoglycan Antibody, clone 9.2.27, clone 9.2.27, Chemicon®, from mouse