MilliporeSigma
  • SARS-CoV-2 Spike protein suppresses CTL-mediated killing by inhibiting immune synapse assembly.
SARS-CoV-2 Spike protein suppresses CTL-mediated killing by inhibiting immune synapse assembly.
The Journal of experimental medicine (2022-11-16)
Anna Onnis, Emanuele Andreano, Chiara Cassioli, Francesca Finetti, Chiara Della Bella, Oskar Staufer, Elisa Pantano, Valentina Abbiento, Giuseppe Marotta, Mario Milco D'Elios, Rino Rappuoli, Cosima T Baldari
ABSTRACT
CTL-mediated killing of virally infected or malignant cells is orchestrated at the immune synapse (IS). We hypothesized that SARS-CoV-2 may target lytic IS assembly to escape elimination. We show that human CD8+ T cells upregulate the expression of ACE2, the Spike receptor, during differentiation to CTLs. CTL preincubation with the Wuhan or Omicron Spike variants inhibits IS assembly and function, as shown by defective synaptic accumulation of TCRs and tyrosine phosphoproteins as well as defective centrosome and lytic granule polarization to the IS, resulting in impaired target cell killing and cytokine production. These defects were reversed by anti-Spike antibodies interfering with ACE2 binding and reproduced by ACE2 engagement by angiotensin II or anti-ACE2 antibodies, but not by the ACE2 product Ang (1-7). IS defects were also observed ex vivo in CTLs from COVID-19 patients. These results highlight a new strategy of immune evasion by SARS-CoV-2 based on the Spike-dependent, ACE2-mediated targeting of the lytic IS to prevent elimination of infected cells.
Sigma-Aldrich
Angiotensin Fragment 1-7 acetate salt hydrate, ≥90% (HPLC), powder
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)