MilliporeSigma
  • High Definition Confocal Imaging Modalities for the Characterization of Tissue-Engineered Substitutes.
High Definition Confocal Imaging Modalities for the Characterization of Tissue-Engineered Substitutes.
Methods in molecular biology (Clifton, N.J.) (2018-04-25)
Dominique Mayrand, Julie Fradette
ABSTRACT
Optimal imaging methods are necessary in order to perform a detailed characterization of thick tissue samples from either native or engineered tissues. Tissue-engineered substitutes are featuring increasing complexity including multiple cell types and capillary-like networks. Therefore, technical approaches allowing the visualization of the inner structural organization and cellular composition of tissues are needed. This chapter describes an optical clearing technique which facilitates the detailed characterization of whole-mount samples from skin and adipose tissues (ex vivo tissues and in vitro tissue-engineered substitutes) when combined with spectral confocal microscopy and quantitative analysis on image renderings.
Sigma-Aldrich
Staurosporine from Streptomyces sp., Molecular Biology, ≥95% (HPLC)
Sigma-Aldrich
Mouse IgG1 Negative Control Antibody, clone 1E2.2, clone 1E2.2, 1 mg/mL, Chemicon®
Sigma-Aldrich
Normal Sheep IgG, Normal Sheep IgG Polyclonal Antibody control validated for use in Western Blotting.