MilliporeSigma
3D Endothelial Cell Migration.
Methods in molecular biology (Clifton, N.J.) (2018-03-12)
Kathryn A Jacobs, Julie Gavard
ABSTRACT
Endothelial cells have the capacity to shift between states of quiescence and angiogenesis. The early stage of angiogenesis, sprouting, occurs with the synchronized activities of tip cells, which lead the migration of the sprout, and stalk cells, which elongate this vessel sprout. Here, we describe a method to study in vitro this early and rapid stage of sprouting angiogenesis.
Sigma-Aldrich
Fibroblast Growth Factor-Basic from bovine pituitary, FGF-Basic, suitable for cell culture