MilliporeSigma
  • Recent Advancement in Boron-Based Efficient and Pure Blue Thermally Activated Delayed Fluorescence Materials for Organic Light-Emitting Diodes.
Recent Advancement in Boron-Based Efficient and Pure Blue Thermally Activated Delayed Fluorescence Materials for Organic Light-Emitting Diodes.
Frontiers in chemistry (2020-06-09)
Hyuna Lee, Durai Karthik, Raju Lampande, Jae Hong Ryu, Jang Hyuk Kwon
ABSTRACT
In the last few years, electron-deficient materials have been actively researched for application in organic light-emitting diode (OLED) as dopant and electron-transporting materials. The boron-containing materials are interesting as they give good emissive properties in solid state with an electron-accepting character. Recently, many boron-containing materials are used as emissive materials for thermally activated delayed fluorescence (TADF) OLED applications. In this review, boron acceptor-based push-pull small molecules used for application in blue TADF OLEDs are reviewed, covering their different types of acceptor, molecular design, structure-property relation, material properties, and device properties. Also, the importance of boron acceptors to address the key issue of blue TADF OLEDs is discussed.
Sigma-Aldrich
t-DABNA, ≥99% (HPLC)