MilliporeSigma
  • DBZ, a CNS-specific DISC1 binding protein, positively regulates oligodendrocyte differentiation.
DBZ, a CNS-specific DISC1 binding protein, positively regulates oligodendrocyte differentiation.
Glia (2014-02-01)
Shoko Shimizu, Yoshihisa Koyama, Tsuyoshi Hattori, Taro Tachibana, Tomohiko Yoshimi, Hisayo Emoto, Yuji Matsumoto, Shingo Miyata, Taiichi Katayama, Akira Ito, Masaya Tohyama
ABSTRACT
Recent studies have shown changes in myelin genes and alterations in white matter structure in a wide range of psychiatric disorders. Here we report that DBZ, a central nervous system (CNS)-specific member of the DISC1 interactome, positively regulates the oligodendrocyte (OL) differentiation in vivo and in vitro. In mouse corpus callosum (CC), DBZ mRNA is expressed in OL lineage cells and expression of DBZ protein peaked before MBP expression. In the CC of DBZ-KO mice, we observed delayed myelination during the early postnatal period. Although the myelination delay was mostly recovered by adulthood, OLs with immature structural features were more abundant in adult DBZ-KO mice than in control mice. DBZ was also transiently upregulated during rat OL differentiation in vitro before myelin marker expression. DBZ knockdown by RNA interference resulted in a decreased expression of myelin-related markers and a low number of cells with mature characteristics, but with no effect on the proliferation of oligodendrocyte precursor cells. We also show that the expression levels of transcription factors having a negative-regulatory role in OL differentiation were upregulated when endogenous DBZ was knocked down. These results strongly indicate that OL differentiation in rodents is regulated by DBZ.
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 4 Antibody, clone 3A11, clone 3A11, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid