MilliporeSigma
  • Loss of RNA-binding motif protein 3 expression is associated with right-sided localization and poor prognosis in colorectal cancer.
Loss of RNA-binding motif protein 3 expression is associated with right-sided localization and poor prognosis in colorectal cancer.
Histopathology (2015-04-30)
Nathaniel Melling, Ronald Simon, Martina Mirlacher, Jakob R Izbicki, Philip Stahl, Luigi M Terracciano, Carsten Bokemeyer, Guido Sauter, Andreas H Marx
ABSTRACT
RNA-binding motif protein 3 (RBM3) has recently been suggested as a prognostic biomarker in an array of human cancers. This study aimed to examine its effects in colorectal cancers. RBM3 expression was analysed by immunohistochemistry on a tissue microarray containing 1800 colorectal cancers (CRCs). Nuclear RBM3 immunohistochemical staining was found in 95.9% of all interpretable CRCs. Loss of RBM3 expression was linked to advanced tumour stage (P < 0.0001), right-sided tumour localization (P < 0.0001), and poor prognosis (P = 0.0003). In a multivariable analysis including RBM3 staining, tumour grade, tumour stage, and nodal status, only tumour stage and nodal status proved to be independent prognostic markers (P < 0.0001 each), whereas the prognostic impact of RBM3 staining was not significant (P = 0.2655). Our observations indicate that loss of RBM3 expression is an unfavourable prognostic marker in CRC, and is linked to right-sided tumour localization.
Sigma-Aldrich
Anti-RBM3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution