MilliporeSigma
Methylated DNA immunoprecipitation (MeDIP).
Methods in molecular biology (Clifton, N.J.) (2008-11-07)
Fabio Mohn, Michael Weber, Dirk Schübeler, Tim-Christoph Roloff
ABSTRACT
Methylated DNA immunoprecipitation (MeDIP) is a versatile immunocapturing approach for unbiased detection of methylated DNA. In brief, genomic DNA is randomly sheared by sonication and immunoprecipitated with a monoclonal antibody that specifically recognizes 5-methylcytidine. The resulting enrichment of methylated DNA in the immunoprecipitated fraction can be determined by PCR to assess the methylation state of individual regions. Alternatively, MeDIP can be combined with large-scale analysis using microarrays as a genome-wide experimental readout. This protocol has been applied to generate comprehensive DNA methylation profiles on a genome-wide scale in mammals and plants, and further to identify abnormally methylated genes in cancer cells.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 90%, powder, white, ~80 U/mg