MilliporeSigma
Functional prediction and characterization of Dip2 gene in mice.
Cell biology international (2019-01-24)
Jun Ma, Li Chen, Xiao-Xiao He, Ya-Jun Wang, Hua-Li Yu, Zi-Xuan He, Lu-Qing Zhang, Yao-Wu Zheng, Xiao-Juan Zhu
ABSTRACT
Disconnected interacting protein 2 (DIP2) is a highly conserved protein family among invertebrates and vertebrates, but its function remains unclear. In this paper, we summarized the conservation of gene sequences and protein domains of DIP2 family members and predicted that they may have a similar functional role in acetyl-coenzyme A (acetyl-CoA) synthesis. We then used the most characterized member, disconnected interacting protein 2 homolog A (DIP2A), for further study. DIP2A is a cytoplasmic protein that is preferentially localized to mitochondria, and its acetyl-CoA synthetase activity has been demonstrated in vitro. Furthermore, the level of acetyl-CoA in HEK293 cells overexpressing DIP2A was increased, which is consistent with its metabolically related function. Together, these data enrich the evolutionary and functional characterization of dip2 genes and provide significant insights into the identification and application of other homologs of DIP2.
Sigma-Aldrich
Anti-DIP2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DIP2C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution