MilliporeSigma
  • Purification and Identification of Natural Inhibitors of Protein Arginine Methyltransferases from Plants.
Purification and Identification of Natural Inhibitors of Protein Arginine Methyltransferases from Plants.
Molecular and cellular biology (2022-03-22)
Zhengxin Wang, Ling Xiong, Quanbo Xiong
ABSTRACT
Protein arginine methyltransferase (PRMT) enzymes catalyze posttranslational modifications of target proteins and are often upregulated in human cancers. In this study, we purified two chemical compounds from seeds of Foeniculum vulgare based on their ability to inhibit the enzymatic activity of PRMT5. These two compounds were identified as Pheophorbide a (PPBa) and Pheophorbide b (PPBb), two breakdown products of chlorophyll. PPBa and PPBb inhibited the enzymatic activity of both Type I and Type II PRMTs with IC50 values at sub micromole concentrations, inhibited the arginine methylation of histones in cells, and suppressed proliferation of prostate cancer cells. Molecular docking results predicted that PPBa binds to an allosteric site in the PRMT5 structure with a high affinity (ΔG = -9.0 kcal/mol) via hydrogen bond, ionic, and π-π stacking interactions with amino acid residues in PRMT5. Another group of natural compounds referred to as protoporphyrins and sharing structural similarity with pheophorbide also inhibited the PRMT enzymatic activity. This study is the first report on the PRMT-inhibitory activity of the tetrapyrrole macrocycles and provides useful information regarding the application of these compounds as natural therapeutic reagents for cancer prevention and treatment.
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, symmetric (SYM10), serum, Upstate®
Sigma-Aldrich
DS-437, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
EPZ015666, ≥98% (HPLC)