MilliporeSigma
  • The severity of behavioural symptoms in FTD is linked to the loss of GABRQ-expressing VENs and pyramidal neurons.
The severity of behavioural symptoms in FTD is linked to the loss of GABRQ-expressing VENs and pyramidal neurons.
Neuropathology and applied neurobiology (2022-02-14)
Priya Gami-Patel, Marta Scarioni, Femke H Bouwman, Baayla D C Boon, John C van Swieten, Netherlands Brain Bank, Annemieke J M Rozemuller, August B Smit, Yolande A L Pijnenburg, Jeroen J M Hoozemans, Anke A Dijkstra
ABSTRACT
The loss of von Economo neurons (VENs) and GABA receptor subunit theta (GABRQ) containing neurons is linked to early changes in social-emotional cognition and is seen in frontotemporal dementia (FTD) due to C9orf72 repeat expansion. We investigate the vulnerability of VENs and GABRQ-expressing neurons in sporadic and genetic forms of FTD with different underlying molecular pathology and their association with the presence and severity of behavioural symptoms. We quantified VENs and GABRQ-immunopositive neurons in the anterior cingulate cortex (ACC) in FTD with underlying TDP43 (FTLD-TDP) (n = 34), tau (FTLD-tau) (n = 24) or FUS (FTLD-FUS) (n = 8) pathology, neurologically healthy controls (n = 12) and Alzheimer's disease (AD) (n = 7). Second, we quantified VENs and the GABRQ-expressing population in relation to presence of behavioural symptoms in the first years of disease onset. The number of VENs and GABRQ-expressing neurons and the ratio of VENs and GABRQ-expressing neurons over total Layer 5 neuronal population decreased in FTLD-TDP and FTLD-FUS, but not in FTLD-tau, compared to control and AD. The severity of early behavioural symptoms in all donors correlated with a lower VEN and GABRQ neuronal count. We show that in FTD, a loss of VENs together with GABRQ-expressing pyramidal neurons is associated with TDP43 and FUS pathology. No significant loss was found in donors with FTLD-tau pathology; however, this could be due to the specific MAPT mutation studied and small sporadic FTLD-tau sample size. Overall, we show the GABRQ-expressing population correlates with behavioural changes and suggest they are key in modulating behaviour in FTD.
Sigma-Aldrich
Anti-GABRE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution