MilliporeSigma
  • Induction of beige-like adipocyte markers and functions in 3T3-L1 cells by Clk1 and PKCβII inhibitory molecules.
Induction of beige-like adipocyte markers and functions in 3T3-L1 cells by Clk1 and PKCβII inhibitory molecules.
Journal of cellular and molecular medicine (2022-07-09)
Achintya Patel, Tradd Dobbins, Xiaoyuan Kong, Rehka Patel, Gay Carter, Linette Harding, Robert P Sparks, Niketa A Patel, Denise R Cooper
ABSTRACT
Excessive dietary intake of fat results in its storage in white adipose tissue (WAT). Energy expenditure through lipid oxidation occurs in brown adipose tissue (BAT). Certain WAT depots can undergo a change termed beiging where markers that BAT express are induced. Little is known about signalling pathways inducing beiging. Here, inhibition of a signalling pathway regulating alternative pre-mRNA splicing is involved in adipocyte beiging. Clk1/2/4 kinases regulate splicing by phosphorylating factors that process pre-mRNA. Clk1 inhibition by TG003 results in beige-like adipocytes highly expressing PGC1α and UCP1. SiRNA for Clk1, 2 and 4, demonstrated that Clk1 depletion increased UCP1 and PGC1α expression, whereas Clk2/4 siRNA did not. TG003-treated adipocytes contained fewer lipid droplets, are smaller, and contain more mitochondria, resulting in proton leak increases. Additionally, inhibition of PKCβII activity, a splice variant regulated by Clk1, increased beiging. PGC1α is a substrate for both Clk1 and PKCβII kinases, and we surmised that inhibition of PGC1α phosphorylation resulted in beiging of adipocytes. We show that TG003 binds Clk1 more than Clk2/4 through direct binding, and PGC1α binds to Clk1 at a site close to TG003. Furthermore, we show that TG003 is highly specific for Clk1 across hundreds of kinases in our activity screen. Hence, Clk1 inhibition becomes a target for induction of beige adipocytes.
Sigma-Aldrich
Anti-CIDE-A Antibody, from rabbit, purified by affinity chromatography