MilliporeSigma
  • Folate Protects Hepatocytes of Hyperhomocysteinemia Mice From Apoptosis via Cystic Fibrosis Transmembrane Conductance Regulator (CFTR)-Activated Endoplasmic Reticulum Stress.
Folate Protects Hepatocytes of Hyperhomocysteinemia Mice From Apoptosis via Cystic Fibrosis Transmembrane Conductance Regulator (CFTR)-Activated Endoplasmic Reticulum Stress.
Journal of cellular biochemistry (2017-02-24)
Anning Yang, Yue Sun, Caiyan Mao, Songhao Yang, Min Huang, Mei Deng, Ning Ding, Xiaoling Yang, Minghao Zhang, Shaoju Jin, Yideng Jiang, Ying Huang
ABSTRACT
Folate deficiency is a known risk factor for liver injury; however, the underlying mechanism remains unclear. In this study, we employed a high homocysteine-induced liver injury model of Apolipoprotein E-deficient (ApoE-/- ) mice fed high-methionine diet and found that high homocysteine induced endoplasmic reticulum (ER) stress and liver cell apoptosis by downregulation of cystic fibrosis transmembrane conductance regulator (CFTR) expression; observations that were attenuated with supplementation of dietary folate. The regulation on CFTR expression was mediated by CFTR promoter methylation and trimethylation of lysine 27 on histone H3 (H3K27me3). Mechanistically, folate inhibited homocysteine-induced CFTR promoter methylation and H3K27me3, which resulted in upregulation of CFTR expression, and reduced ER stress and liver cell apoptosis. Further study showed that folate inhibited the expression of DNA methyltransferase 1 and enhancer of zeste homolog 2, downregulated the cellular concentrations of S-adenosylmethionine (SAM) and S-adenosylhomocysteine (SAH) and upregulated the SAM/SAH ratio, leading to the inhibition of Hcy-induced DNA hypermethylation and H3K27me3 in CFTR promoter. In conclusion, our results provide insight into the protective role of folate in homocysteine-induced ER stress and liver cell apoptosis through the regulation of CFTR expression. J. Cell. Biochem. 118: 2921-2932, 2017. © 2017 Wiley Periodicals, Inc. Folate protects hepatocytes of hyperhomocysteinemia mice from apoptosis. Folate alleviates Hcy-induced hepatocyte apoptosis. Folate inhibits Hcy-induced ER stress via upregulation of CFTR expression in hepatocytes. Folate inhibits Hcy-induced methylation of CFTR promotor and H3K27me3.
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-PERK Antibody, clone 4F6.1, clone 4F6.1, from mouse